Micro Eyelash, Ted Pella
Supplier: TED PELLA INC MS
113
MSPP-113EA 19.8 USD
MSPP-113
Micro Eyelash, Ted Pella
Microtomes and Microtome Assemblies Microtome Blades and Handles
No. 1 superfine eyelash is attached to a fine wooden handle for delicate manipulation of ultra-thin sections in the knife boat of an ultramicrotome or wherever delicate teasing or manipulation is required.
  • Excellent taper and strength