Stainless Steel Beads, Biospec Products
Supplier: BIOSPEC PRODUCTS MS
11079132SS
MSPP-11079132SEA 59.23 USD
MSPP-11079132S
Stainless Steel Beads, Biospec Products
Beads Metallic Beads
Stainless steel beads have a density of 7.9 g/cc. They are used mostly for grinding leaves and seeds. They are made of Type 316 stainless steel - the most corrosion resistant available.
  • Cleaned and reused
  • More economic and time-saving