BeadBeater Laboratory Homogenizer, Biospec Products
Supplier: BIOSPEC PRODUCTS MS
1107900
MSPP-1107900EA 893.81 USD
MSPP-1107900
BeadBeater Laboratory Homogenizer, Biospec Products
Homogenizers Benchtop Homogenizers
The BeadBeater is the ultimate laboratory-scale homogenizer for disrupting aqueous suspensions of yeast, fungi or bacteria. Complete cell lysis of 10 to 80 g (wet weight) of cells is achieved in about three minutes of operation.
  • Ideal for isolating proteins, enzymes and intact intracellular organelles
  • Sample cooling during homogenisation is simple and efficient
  • Powerful, high speed motor
  • Only durable, non-toxic components will be contact the product
  • Designed for batches up to 80 g (wet weight) in 200 ml of medium
  • Highly efficient disruption in shortest possible times
  • Homogeniser clean-up and mantinance is uncomplicated and easy
Delivery information: The BeadBeater is supplied with a 350 ml polycarbonate chamber, rotor assembly, motor base, ice-water cooling jacket, and one pound of glass beads of your choice.
Caution: Glass beads and zirconia-silica beads having diameters of 0.1 mm, 0.5 mm and 1 mm are the prefered grinding media. Larger and heavier beads such as steel beads cannot be adequately agitated.