RNaseAlert® Nuclease Control Reagents, Integrated DNA Technologies
Supplier: 0000014838
11-02-01-02
MSPP-11020102EA 192.31 USD
MSPP-11020102
RNaseAlert® Nuclease Control Reagents, Integrated DNA Technologies
Nucleic Acid Reagents Primers and Probes
These reagents are fluorescence-quenched oligonucleotide probes that emit a fluorescence signal only after nuclease degradation.
  • Used for qualitative detection of nuclease contamination
  • Used as quantitative tool to study enzyme kinetics
  • Employs a FAM™ reporter
The assay can be read visually or measured and quantified using fluorometry.
Delivery information: It includes single use tubes, buffer, water, enzyme, and nuclease decontamination solution.