Hose Clip, ibidi
Supplier: 0000027849
10821
MSPP-10821PK 192.31 USD
MSPP-10821
Hose Clip, ibidi
Clamps Tubing and Sanitary Clamps
A hose clip used for temporary or permanent closing of silicone tubes.
  • Allows easy closing and re-opening of silicone tubing
  • Delivered as part of the Fluidic Unit
  • Temperature stability: Up to 80 °C (not autoclavable)
Ideal for all flow applications.