In-line Luer Injection Port, ibidi
Supplier: 0000027849
10820
MSPP-10820PK 192.31 USD
MSPP-10820
In-line Luer Injection Port, ibidi
Catheters
An in-line injection port used to inject liquids into a running flow experiment.
  • Allows fluids to be injected with a needle into a running experiment without stopping the flow
  • Silicone septum, individually packed, sterilized
  • Temperature stability: Up to 90 °C (not autoclavable)
This in-line injection port is adaptable to a tubing of ID 0.5 - 1.6 mm (ie. Ibidi perfusion sets) when combined with a Luer Lock Connector Male and a Luer Lock Connector Female.
Max. pressure: 500 mbar