Coverslips for Sticky-Slides, Ibidi
Supplier: 0000027849
10812 10813
MSPP-10812PK 423.08 USD
MSPP-10812 MSPP-10813
Coverslips for Sticky-Slides, Ibidi
Cover Glasses
Coverslips for ibidi sticky-slides, available in glass or polymer formats.
  • Polymer coverslip available with ibiTreat tissue culture treatment, or uncoated
  • Glass coverslip made from high-quality D 263 M Schott glass
Coverslips for sticky-Slides, designed for high–end microscopic analysis with uncompromised resolution and choice of wavelength.