Coverslips for Chamber Slides, Removable, Ibidi
Supplier: IBIDI USA MS
10811
MSPP-10811PK 153.85 USD
MSPP-10811
Coverslips for Chamber Slides, Removable, Ibidi
Cover Glasses
Glass coverslips for 'ibidi Chamber, removable' slides.
  • Ideal for immunofluorescence assays
  • Compatible with the 3-, 8-, and 12-chamber formats
  • Compatible with all mounting media
  • No autofluorescence
  • Low birefringence
Certifications: In compliance with DIN ISO 4796-1 standards.