EasySep™ Human NK Cell Isolation Kits
Supplier: STEMCELL Technologies
17955 17955RF 100-0960
MSPP-17955EA 1466.15 USD
MSPP-17955 MSPP-17955RF MSPP-100-0960
EasySep™ Human NK Cell Isolation Kits
Cell Separation Media
Immunomagnetic isolation of untouched human NK cells
  • Fast, easy-to-use and column-free
  • Up to 94% purity with high recovery
  • Isolated cells are untouched
The EasySep™ Human NK Cell Isolation Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection. The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles. The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube. This product can be used in place of the EasySep™ Human NK Cell Enrichment Kit (Catalog #19055) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').