BloodStor® 55-5 Freezing Medium
Supplier: STEMCELL Technologies
07937
MSPP-07937EA 777.54 USD
MSPP-07937
BloodStor® 55-5 Freezing Medium
Cell Freezing Media
Optimized biopreservation reagent for hematopoietic cells and tissues.
  • Ready-to-use
  • Animal component-free
  • Sterile, USP vial
  • cGMP manufactured with USP grade/highest-quality components
  • Sterility, endotoxin, and cell-based release testing
Cryopreserve stem cells and other cells isolated from umbilical cord blood, peripheral blood, bone marrow, and other biologics with BloodStor® 55-5, an animal component-free and ready-to-use medium. cGMP-manufactured with USP-grade components, BloodStor® 55-5 is sterile and tested for cell-based release and endotoxins. BloodStor® 55-5 is pre-formulated with 55% (w/v) DMSO USP, 5% (w/v) Dextran-40 USP, and WFI-quality water. It is available in a variety of formats.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').