Human Pluripotent Stem Cell Trilineage Differentiation qPCR Array
Supplier: STEMCELL Technologies
07515
MSPP-07515EA 685.46 USD
MSPP-07515
Human Pluripotent Stem Cell Trilineage Differentiation qPCR Array
Assays Nucleic Acid Assays
For characterization of hPSC trilineage differentiation to endoderm, mesoderm, and ectoderm germ layers.
The Human Pluripotent Stem Cell (hPSC) Trilineage Differentiation Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of hPSCs and their trilineage differentiation capacity. hPSCs, including embryonic stem (ES) and induced pluripotent stem (iPS) cells, are self-renewing and have the ability to differentiate into cells of the three embryonic germ layers: ectoderm, mesoderm, and endoderm. This qPCR array provides the gene expression profile of undifferentiated ES and iPS cells and their trilineage derivatives following in vitro directed or spontaneous differentiation, thereby validating the ability of a cell line to differentiate to the three germ layers. Genes were selected based on their demonstrated differential expression in ES and iPS cells compared with hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (Adewumi et al.; Bock et al.).
qPCR is used to determine changes in steady-state mRNA levels of gene expression across multiple samples, generally normalized to the relative expression of internal control genes. Gene-specific primers amplify target sequences within cDNA pools reverse-transcribed from mRNA and, as with TaqMan® technology, hybridized sequence-specific probes provide a fluorescent signal from the 5' exonuclease activity of Taq DNA polymerase. The accumulation rate of the fluorescent signal is used to quantify the sample cDNA, and thereby the amount of mRNA in the original cell lysate. This qPCR array contains validated primers and probes for detection of 90 genes whose expression is correlated with undifferentiated hPSCs or their derivatives undergoing the early stages of differentiation, as well as six endogenous (housekeeping) control genes. TBP (TATA box-binding protein) qPCR Control Template is provided as a synthetic DNA positive control.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').