Collagenase Type V
Supplier: STEMCELL Technologies
07430 07431 100-0681
MSPP-07430EA 156.38 USD
MSPP-07430 MSPP-07431 MSPP-100-0681
Collagenase Type V
Cell Culture Supplements and Additives
For digestion of native collagen fibrils.
  • High levels of collagenase and caseinase activities
Digest native collagen fibrils in connective tissues with Collagenase Type V, sourced from Clostridium histolyticum. Collagenase preparations typically contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). This Collagenase Type V preparation contains high levels of collagenase and caseinase activities.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').