Anti-Adherence Rinsing Solution
Supplier: STEMCELL Technologies
07010
MSPP-07010EA 86.23 USD
MSPP-07010
Anti-Adherence Rinsing Solution
Cell Separation Media
Anti-Adherence Rinsing Solution is a surfactant solution for pre-treating cultureware to reduce surface tension and prevent cell adhesion. It can be used with plates, flasks, pipettes, strainers and other cultureware.
Adherence rinsing solution is required for use with the following:
- AggreWell™ plates (e.g. Catalog MSPP-34811)
- Organoid growth media, including HepatiCult™ (Catalog MSPP-06030)
- PancreaCult™ (Catalog MSPP-06040)
Species: Human, mouse, non human primate, rat and others.
Caution: Note: Formerly known as AggreWell™ Rinsing Solution, name change effective Dec 2017.
Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet ( collectively, the ‘Product Restrictions’).