HepatiCult™ Organoid Growth Medium (Mouse)
Supplier: STEMCELL Technologies
06030
MSPP-06030EA 610.77 USD
MSPP-06030
HepatiCult™ Organoid Growth Medium (Mouse)
Cell Culture Media
Cell culture medium for establishment and maintenance of mouse hepatic progenitor organoids.
HepatiCult™ Organoid Growth Medium (Mouse) is a serum-free, defined cell culture medium for establishment and maintenance of mouse hepatic progenitor organoids. These organoids, or 'mini-livers', provide an in vitro organotypic culture system for studying hepatic stem and progenitor cells. Organoids grown in HepatiCult™ feature an epithelium expressing genes marking hepatic stem and progenitor cells (PROM1, AXIN2, SOX9 and CD44), ducts (KRT19 and HNF1b) and hepatocytes (HNF4a, AFP).
Hepatic organoids can be passaged every 4 to 7 days, can be cryopreserved, and are primed for downstream differentiation. HepatiCult™ supports mouse hepatic organoid culture either embedded in Corning® Matrigel® domes or in a dilute Matrigel® suspension. Organoid culture enables convenient in vitro characterization of the hepatic epithelium in a physiologically relevant system and reduces the need for animal use.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').