MesenCult™ Osteogenic Differentiation Kit (Human)
Supplier: STEMCELL Technologies
5465
MSPP-05465EA 566.15 USD
MSPP-05465
MesenCult™ Osteogenic Differentiation Kit (Human)
Cell Culture Media
For the in vitro differentiation of human MSCs into osteoblasts.
MesenCult™ Osteogenic Differentiation Kit (Human) is specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (also known as mesenchymal stem cells or MSCs) into cells of the osteogenic lineage. This kit is suitable for the differentiation of human bone marrow (BM)- or adipose-derived MSCs previously culture-expanded in serum-containing medium (e.g. MesenCult™ Proliferation Kit or MesenCult™-hPL Medium) or animal component-free MesenCult™-ACF Plus Medium.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').