Mouse IL-5 ELISA Kit
Supplier: STEMCELL Technologies
02024
MSPP-02024EA 1134.15 USD
MSPP-02024
Mouse IL-5 ELISA Kit
Assays ELISAs
For detection and measurement of mouse interleukin 5.
The Mouse Interleukin 5 (IL-5) ELISA Kit is designed for the quantitative detection and measurement of mouse IL-5 in biological fluids such as serum, plasma, and cell culture supernatants. IL-5 is produced by Th2 cells, eosinophils, and activated mast cells. IL-5 plays an important role in the recruitment, activation, proliferation, and survival of eosinophils.
The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine. The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody.
Addition of the chromogenic enzyme substrate 3,3’, 5,5’ tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample.
The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').