Human IL-23 ELISA Kit
Supplier: STEMCELL Technologies
02016
MSPP-02016EA 1460.08 USD
MSPP-02016
Human IL-23 ELISA Kit
Assays ELISAs
For detection and measurement of human interleukin 23.
The Human Interleukin 23 (IL-23) ELISA Kit is designed for the quantitative detection and measurement of human IL-23 in biological fluids such as serum, plasma, and cell culture supernatants. IL-23 is a heterodimeric cytokine composed of a p19 and a p40 subunit; the p40 subunit is shared with IL-12. IL-23 is secreted by activated macrophages and dendritic cells, and induces the proliferation of memory T cells and the production of IL-17.
The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine. The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample. The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analyzed in parallel.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').