VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
20721103TU
MP97058-166EA 1130.5 USD
MP97058-166
VACUUBRAND® ME 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Pumps Vacuum Diaphragm Pumps
The ME 1C diaphragm vacuum pumps offer a compact, high performance and easy-to-use solution which is perfect for both single and multiple filtrations, solid phase extraction (SPE) and other fluid movement tasks.
  • Excellent for filtration, degassing and SPE
  • Quiet, low vibration
  • Typical service intervals >15000 operating hours
  • Optional flow valve/manometer
Corrosion resistant design, the ME 1C features a robust PTFE diaphragms and valves for excellent corrosion resistance in routine biology and chemistry lab applications. The ME 1C incorporates a PTFE head to provide a full fluoropolymer flow path for maximum chemical resistance to the most challenging applications, including acidic vapors.
Quiet and reliable; these pumps offer whisper-quiet operation, low vibration, and long service intervals; 15000 operating hours in most applications.
The functional, space saving and innovative design with readily accessible top mounted power switch ensures convenient and quick operation.
Certifications:
cTŰVus.
Ordering information:
Two-year warranty.
Delivery information:
Pumps are supplied with 100 to 120 V, 50/60 Hz motors, US plug, operating manual.