VACUUBRAND® RZ 2.5 Rotary Vane Pump, Trade-Up Promotion
Supplier: Brandtech
20798126TU
MP77322-838EA 2750 USD
MP77322-838
VACUUBRAND® RZ 2.5 Rotary Vane Pump, Trade-Up Promotion
Pumps Vacuum Rotary Vane Pumps
The RZ 2.5 from VACUUBRAND® delivers impressive performances in a compact design.
  • Powerful ultimate vacuum even with gas ballast open
  • High-performing, energy efficient motor
  • Whisper quiet with minimal vibrations
  • Small footprint and easy maintenance
The two-stage RZ 2.5 rotary vacuum pump is the optimal choice for any laboratory process demanding low ultimate vacuum at a medium gas flow rate.
The compact, telescopic design makes required maintenance effortless, and the large oil volume capacity provides extended intervals between oil changes.
Common applications include lyophilization, fine vacuum distillation, and evacuation of small volumes.
Certifications:
cTŰVus.
Ordering information:
Two-year warranty.
Delivery information:
Includes pump, operating manual, and US plug.