VACUUBRAND® MD 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Supplier: Brandtech
20696603TU
MP76529-726EA 3320.25 USD
MP76529-726
VACUUBRAND® MD 1C Oil-Free Chemistry Diaphragm Pump, Trade-Up Promotion
Pumps Vacuum Diaphragm Pumps
Powerful 2 millibar performance for evaporation of high boiling point solvents.
  • Oil-free
  • Corrosion-resistant fluoropolymer/borosilicate construction
  • Whisper-quiet
  • Ultra-low maintenance (>15000 service hours)
  • Compact footprint
The MD 1C chemistry diaphragm vacuum pump provides high flow rates at working vacuum levels to evaporate high boiling point solvents.
A three-stage design, with integrated gas ballast provides a high tolerance for condensable vapors. A full fluoropolymer flow path provides superior resistance to chemicals. Pump is oil-free, with typical service intervals in excess of 15000 hours of operation (15 months of 24/7 operation).
Suitable for many benchtop applications, such as rotary evaporation, gel drying, and small vacuum ovens.
Certifications:
cTŰVus.
Ordering information:
Two-year warranty.
Delivery information:
Pumps are supplied with 100 to 120 V, 50/60 Hz motors, US plug, operating manual.