Toluene ≥99.8% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methylbenzene, Methacide, Benzene, methyl, Phenylmethane, Methane, phenyl-, Toluol, FM approval, Methylbenzol

Management of Change (MOC) category = N

V560-10
MKV56010CS 977.65 USD
MKV56010
Toluene ≥99.8% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Toluene
Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅CH₃) (by GC, corrected for water) ≥99.8 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0002 %
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Water (H₂O)(by KF, coulometric) ≤0.02 %
ACS - Sulfur Compounds (as S) ≤0.003 %
Optical Absorbance (1-cm path vs water) - 285 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 286 nm ≤0.70
Optical Absorbance (1-cm path vs water) - 288 nm ≤0.40
Optical Absorbance (1-cm path vs water) - 293 nm ≤0.20
Optical Absorbance (1-cm path vs water) - 300 nm ≤0.10
Optical Absorbance (1-cm path vs water) - 310 nm ≤0.05
Optical Absorbance (1-cm path vs water) - 335 nm ≤0.02
Optical Absorbance (1-cm path vs water) - 400 - 350 nm ≤0.01
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L) ≤10
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR