Tetrahydrofuran ≥99.8%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

Management of Change (MOC) category = N

V558-10
MKV55810CS 2151.14 USD
MKV55810
Tetrahydrofuran ≥99.8%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Tetrahydrofuran
Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₈O) (by GC, corrected for water) ≥99.8 %
Color (APHA) ≤10
Peroxide (as H₂O₂)(at time of packaging) ≤0.005 %
Residue after Evaporation ≤2.0 ppm
Water (H₂O)(by KF, coulometric) ≤0.02 %
Optical Absorbance (1-cm path vs water) - 212 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 230 nm ≤0.40
Optical Absorbance (1-cm path vs water) - 254 nm ≤0.15
Optical Absorbance (1-cm path vs water) - 280 nm ≤0.03
Optical Absorbance (1-cm path vs water) - 320 - 400 nm ≤0.005
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR