Ethyl acetate ≥99.6%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

Management of Change (MOC) category = N

V553-10
MKV55310CS 2266.97 USD
MKV55310
Ethyl acetate ≥99.6%, UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COOCH₂CH₃) (by GC, corrected for water) ≥99.6 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤1 ppm
ACS - Titrable Acid (µeq/g) ≤0.8
ACS - Water (H₂O)(by KF, coulometric) ≤0.04 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 257 nm ≤0.50
Ultraviolet Absorbance (1.00-cm cell vs. water) - 263 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm ≤0.01
Substances Darkened by H₂SO₄ Passes Test
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L) ≤10 ppb
FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L) ≤5 ppb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR