Cyclohexane ≥99% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

V552-10
MKV55210CS 2055.49 USD
MKV55210
Cyclohexane ≥99% (by GC, corrected for water content), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Cyclohexane

Suitable for Liquid Chromatography, Extract/Conc, UV-Spectrophotometry, Organic Residue Analysis

Formula: C₆H₁₂
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
ACS - Assay (C₆H₁₂) (by GC, corrected for water) ≥ 99.0 %
ACS - Color (APHA) ≤ 10
ACS - Residue after Evaporation ≤ 2 ppm
ACS - Substances Darkened by H₂SO₄ Passes Test
ACS - Water (H₂O)(by KF, coulometric) ≤ 0.01 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm ≤ 1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤ 0.60
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤ 0.25
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤ 0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤ 0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤ 0.03
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤ 0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 - 400 nm ≤ 0.01
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤ 1.0 ppb
ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L) ≤ 10 ppb
Trace Impurities - GC-FID Sensitive Impurities (as 2-Octanol) Single Peak (µg/L) ≤ 10 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR