Acetic acid glacial ≥99.7%, ChromAR® for liquid chromatography, for TLC, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

V155-06
MKV15506CS 796.12 USD
MKV15506
Acetic acid glacial ≥99.7%, ChromAR® for liquid chromatography, for TLC, Macron Fine Chemicals™
Acetic acid
Formula: H₃CCOOH
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
Assay (by GC, corrected for water) ≥99.7 %
Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
Chloride (Cl) ≤0.4 ppm
Color (APHA) ≤10
Dilution Test Passes Test
Heavy Metals (by ICP-OES) ≤5 ppm
Identification Passes Test
Residue after Evaporation ≤5 ppm
Substances Reducing Dichromate Passes Test
Substances Reducing Permanganate Passes Test
Sulfate (SO₄) ≤0.4 ppm
Titrable Base (meq/g) ≤0.0004
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.02
Trace Impurities - Copper (Cu) ≤0.10 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm
Trace Impurities - Nickel (Ni) ≤0.1 ppm
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR