Total base number titration solvent mixture ASTM D4739, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

V068-10
MKV06810CS 886.26 USD
MKV06810 JTV068-24
Total base number titration solvent mixture ASTM D4739, Macron Fine Chemicals™
Total base number titration solvent mixture

For ASTM D4739.

UN: 1992
ADR: 3 (6.1),II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Density (g/mL) at 25°C 1.030 - 1.040
Water (v/v) 0.9 - 1.1 %
Assay (by GLC) - Isopropanol (v/v) 32 - 34 %
Assay (by GLC) - Toluene (v/v) 32 - 34 %
Assay (by GLC) - Chloroform (v/v) 32 - 34 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR