2-Propanol ≥99.0% Lab Grade for histology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

Management of Change (MOC) category = N

MACRH604-26
JTH604-26CS 2363.58 USD
JTH604-26 MKH604-26
2-Propanol ≥99.0% Lab Grade for histology, Macron Fine Chemicals™
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Flash Pt: 12.0 °C
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃CHOHCH₃) ≥99.0 %
Color (APHA) ≤10
Density (g/mL) at 25°C 0.781 - 0.783
Residue after Evaporation ≤0.001 %
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Appearance (clear, colorless liquid free from foreign matter)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR