Hexane (mixture of isomers) ≥99.5% (by GC, sum of isomers + methylcyclopentane) (95% n-Hexane), UltimAR® for liquid chromatography, for organic residue analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hexanes, Isohexane

Management of Change (MOC) category = NI

H487-23 H487-06 H487-10
MKH48723EA 1266.99 USD
MKH48723 MKH48706 MKH48710 JTH487-62
Hexane (mixture of isomers) ≥99.5% (by GC, sum of isomers + methylcyclopentane) (95% n-Hexane), UltimAR® for liquid chromatography, for organic residue analysis, Macron Fine Chemicals™
Hexane (mixture of isomers)

Suitable for Liquid Chromatography, Extract/Conc, UV-Spectrophotometry, Organic Residue Analysis


UltimAR®—Solvents that meet ACS specifications for liquid chromatography, UV-spectrophotometry, and general purpose use.

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
methylcyclopentane)(by GC) ≥99.5 %
ACS - Assay (sum of 5 isomers, total hexanes plus ≥99.5 %
ACS - Assay (as n-hexane) (by GC, corrected for water) ≥95.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0001 %
ACS - Sulfur Compounds (as S) ≤0.005 %
ACS - Thiophene Passes Test
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.01 %
ACS - Water-Soluble Titrable Acid (meq/g) ≤0.0003
Ultraviolet Absorbance (1.00-cm cell vs. water) - 195 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.07
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 - 400 nm ≤0.005
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L) ≤10 ppb
FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L) ≤5 ppb
FID - Sum of the Peaks (µg/L) Pass
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤5
Neat Solvent Front Characterization - Largest Peak, 0-0.4 x RT (ng/mL)
Filtered through a 0.2 micron filter.
Packaged under Nitrogen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR