Chloroform ≥99.9% stabilized, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Management of Change (MOC) category = N

H407-10
MKH40710CS 1357.47 USD
MKH40710
Chloroform ≥99.9% stabilized, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Chloroform

For HPLC. Stabilized with 1% ethanol. Filtered through a 0.2µm element.

Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CHCl₃ including stabilizer) (by GC) ≥99.9 %
ACS - Residue after Evaporation ≤0.0003 %
ACS - Water (H₂O)(by KF, coulometric) ≤0.03 %
Optical Absorbance (1-cm path vs water) - 245 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 255 nm ≤0.15
Optical Absorbance (1-cm path vs water) - 260 nm ≤0.05
Optical Absorbance (1-cm path vs water) - 270 nm ≤0.02
Optical Absorbance (1-cm path vs water) - 290 - 400 nm ≤0.01
Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
Acid and Chloride Passes Test
Color (APHA) ≤10
Free Chlorine Passes Test
Lead (Pb) ≤0.05 ppm
Substances Darkened by H₂SO₄ Passes Test
Optical Absorbance (1-cm path vs water) - Suitability for Use in Dithizone Tests Passes Test
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR