Sodium thiosulfate 0.01 N (N/100), StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

H362-07 H362-05
MKH36207CS 472.19 USD
MKH36207 MKH36205
Sodium thiosulfate 0.01 N (N/100), StandARd® volumetric solution, Macron Fine Chemicals™
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0095 - 0.0105


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR