Phenolphthalein 1,0% in 95% alcohol, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

Management of Change (MOC) category = N

H306-01 H306-05
MKH30601CS 357.36 USD
MKH30601 MKH30605
Phenolphthalein 1,0% in 95% alcohol, StandARd®, Macron Fine Chemicals™
Phenolphthalein

pH 8.0 to 10.0

Formula: C₂₀H₁₄O₄
Boiling Pt: ∼78 °C (1013 hPa)
Density: ∼0.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
pH ≥10.0
Product Information (not specifications): Appearance (clear, colorless
solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR