Phenolphthalein 1,0% in 95% alcohol, StandARd®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Management of Change (MOC) category = N
H306-01
H306-05
MKH30601CS
357.36
USD
MKH30601
MKH30605
Phenolphthalein 1,0% in 95% alcohol, StandARd®, Macron Fine Chemicals™
Phenolphthalein
pH 8.0 to 10.0
Formula:
C₂₀H₁₄O₄ Boiling Pt: ∼78 °C (1013 hPa) Density: ∼0.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 UN: 1170 ADR: 3,II |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
pH | ≥10.0 |
Product Information (not specifications): Appearance (clear, colorless | |
solution) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...