Nessler reagent, StandARd® for ammonia nitrogen determination, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
Management of Change (MOC) category = N
H261-01
H261-07
MKH26101CS
574.14
USD
MKH26101
MKH26107
Nessler reagent, StandARd® for ammonia nitrogen determination, Macron Fine Chemicals™
Nessler's reagent
For nitrogen in ammonia.
Boiling Pt:
356.6 °C (1013 hPa) Melting Pt: –38.9 °C Density: 13.6 g/cm³ (20 °C) |
MDL Number:
MFCD00049656 CAS Number: 7783-33-7 EINECS: 231-990-4 UN: 3289 ADR: 6.1 (8),II Merck Index: 12,07862 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Suitability | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...