1-Octanesulfonic acid sodium salt, monohydrate ≥98%, OR, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Sodium 1-octansulfonate, Sodium octane-1-sulphonate

Management of Change (MOC) category = N

H165-56
MKH16556EA 958.98 USD
MKH16556
1-Octanesulfonic acid sodium salt, monohydrate ≥98%, OR, Macron Fine Chemicals™
1-Octanesulphonic acid sodium salt

Powder. For use in PIC and HPLC applications.

Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃(CH₂)₇SO₃Na·H₂O) ≥98.0 %
Solubility Passes Test
Optical Density at 210 nm (au) ≤0.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR