EDTA disodium salt 0.0575 M in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

Management of Change (MOC) category = NI

H109-05
MKH10905CS 388.42 USD
MKH10905
EDTA disodium salt 0.0575 M in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Molarity 0.0570 - 0.0580

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR