Sodium hexametaphosphate, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

E024-61
MKE02461EA 149.23 USD
MKE02461
Sodium hexametaphosphate, Macron Fine Chemicals™
Sodium hexametaphosphate

Crystals.

Formula: Na₆P₆O₁₈
Melting Pt: 610 °C
Density: 1.25 g/cm³ (20 °C)
MDL Number: MFCD00136045
CAS Number: 10124-56-8
EINECS: 233-343-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Phosphorus Pentoxide (P₂O₅) ≥65.0 %
pH of 1% Solution at 25°C 6.7 - 7.2
Appearance (clear, glass-like crystals)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR