Magnesium sulfate, anhydrous ≥99.0%, powder USP, Macron Fine Chemicals™

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

Management of Change (MOC) category = R

BS15-11
JTBS15-10EA 1477.97 USD
JTBS15-10 JTBS15-11
Magnesium sulfate, anhydrous ≥99.0%, powder USP, Macron Fine Chemicals™
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
MDL Number: MFCD00011110
CAS Number: 7487-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Appearance Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Loss on Drying at 105°C ≤ 2 %
Solubility Passes Test
USP - pH (1 in 20) 5.0 - 9.2
USP - Chloride (Cl) ≤ 0.014 %
USP - Selenium (Se) ≤ 0.003 %
USP - Iron (Fe) ≤ 20 ppm
USP - Assay (as MgSO₄) (ignited basis) ≥ 99.0 %
Thru U.S. No. 100 Sieve ≥ 85 %
Cadmium (Cd), For Information Only
Cobalt (Co), For Information Only
Nickel (Ni), For Information Only
Vanadium (V), For Information Only
Selenium (Se), For Information Only
Chromium (Cr), For Information Only
Copper (Cu), For Information Only
Molybdenum (Mo), For Information Only
Manganese (Mn), For Information Only
Appearance (fine, white powder)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR