Methanol ≥99.5% (by GC), Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

MACR8820-06 MACR8820-28
MK882006CS 507.13 USD
MK882006 MK8820-28
Methanol ≥99.5% (by GC), Macron Fine Chemicals™
Methanol
Formula: CH₃OH
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
NF - Assay (CH₃OH) (by GC) ≥99.5 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Acidity Passes Test
NF - Alkalinity (as NH₃) ≤3 ppm
NF - Nonvolatile Residue ≤0.001 %
NF - Readily Carbonizable Substances Passes Test
NF - Readily Oxidizable Substances Passes Test
NF - Acetone, Aldehydes (as (CH₃)₂CO) ≤0.003 %
NF - Water (H₂O) ≤0.1 %
Appearance (clear, colorless liquid) No Class 1 or Class 2 Residual
Solvents are present in this product. Only Class 3 Solvents are likely
to be present. Residual concentration of all Class 3 Solvents is
<0.5%.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR