Acetic acid glacial 99.5-100.5%, AR Select® ACS for trace analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

Management of Change (MOC) category = N

8817-14
MK881714CS 763.25 USD
MK881714
Acetic acid glacial 99.5-100.5%, AR Select® ACS for trace analysis, Macron Fine Chemicals™
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
ACS - Assay (CH₃COOH) 99.5 - 100.5 %
ACS - Assay (by GC, corrected for water) ≥ 99.7 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤ 0.01 %
ACS - Arsenic and Antimony (as As) ≤ 0.005 ppm
ACS - Chloride (Cl) ≤ 0.0001 %
ACS - Color (APHA) ≤ 10
Dilution Test Passes Test
ACS - Heavy Metals (by ICP-OES) ≤ 5 ppm
ACS - Residue after Evaporation ≤ 0.001 %
ACS - Substances Reducing Dichromate Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Sulfate (SO₄) ≤ 0.0001 %
ACS - Titrable Base (meq/g) ≤ 0.0004
Determine by Flame Photometry & ICP - Aluminum (Al) ≤ 100 ppb
Determine by Flame Photometry & ICP - Barium (Ba) ≤ 100 ppb
Determine by Flame Photometry & ICP - Beryllium (Be) ≤ 10 ppb
Determine by Flame Photometry & ICP - Bismuth (Bi) ≤ 10 ppb
Determine by Flame Photometry & ICP - Boron (B) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Cadmium (Cd) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Calcium (Ca) ≤ 500.0 ppb
Determine by Flame Photometry & ICP - Chromium (Cr) ≤ 100 ppb
Determine by Flame Photometry & ICP - Cobalt (Co) ≤ 50 ppb
Determine by Flame Photometry & ICP - Copper (Cu) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Gallium (Ga) ≤ 500 ppb
Determine by Flame Photometry & ICP - Germanium (Ge) ≤ 500 ppb
Determine by Flame Photometry & ICP - Gold (Au) ≤ 500 ppb
Determine by Flame Photometry & ICP - Iron (Fe) ≤ 100 ppb
Determine by Flame Photometry & ICP - Lead (Pb) ≤ 500.0 ppb
Determine by Flame Photometry & ICP - Lithium (Li) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Magnesium (Mg) ≤ 100 ppb
Determine by Flame Photometry & ICP - Manganese (Mn) ≤ 50 ppb
Determine by Flame Photometry & ICP - Mercury (Hg) ≤ 500.0 ppb
Determine by Flame Photometry & ICP - Molybdenum (Mo) ≤ 10 ppb
Determine by Flame Photometry & ICP - Nickel (Ni) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Potassium (K) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Silicon (Si) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Silver (Ag) ≤ 50.0 ppb
Determine by Flame Photometry & ICP - Sodium (Na) ≤ 500.0 ppb
Determine by Flame Photometry & ICP - Strontium (Sr) ≤ 100 ppb
Determine by Flame Photometry & ICP - Thallium (Tl) ≤ 50 ppb
Determine by Flame Photometry & ICP - Tin (Sn) ≤ 100.0 ppb
Determine by Flame Photometry & ICP - Vanadium (V) ≤ 10 ppb
Determine by Flame Photometry & ICP - Zinc (Zn) ≤ 100 ppb
Determine by Flame Photometry & ICP - Zirconium (Zr) ≤ 10 ppb
Determine by Flame Photometry & ICP
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR