Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

MACR8814-28
JT8814-9SCS 1317.93 USD
JT8814-9S JT8814-QM MK8814-28
Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™
Methanol
Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (CH₃OH) (by GC) ≥99.5 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Acidity Passes Test
NF - Alkalinity (as NH₃) ≤3 ppm
NF - Nonvolatile Residue ≤0.001 %
NF - Readily Carbonizable Substances Passes Test
NF - Readily Oxidizable Substances Passes Test
NF - Acetone, Aldehydes (as (CH₃)₂CO) ≤0.003 %
NF - Water (H₂O) ≤0.1 %
Appearance (clear, colorless liquid)
No Class 1 or Class 2 Residual Solvents are present in this product.
Only Class 3 Solvents are likely to be present. Residual concentration of all Class 3 Solvents is <0.5%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR