Xylene (mixture of isomers) ≥98.5% (by GC) for trace metal analysis, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

8802-04
MK880204CS 1483.8 USD
MK880204
Xylene (mixture of isomers) ≥98.5% (by GC) for trace metal analysis, Macron Fine Chemicals™
Xylene (mixture of isomers)

For use as diluent for metallo-organic standards.

Formula: C₆H₄(CH₃)₂
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (m,p,o-xylenes and ethylbenzene) (by GC) ≥ 98.5 %
Acidity (µeq/g) ≤ 0.3
Arsenic and Antimony (as As) ≤ 0.010 ppm
Chloride (Cl) ≤ 3 ppm
Color (APHA) ≤ 10
Heavy Metals (as Pb) ≤ 1 ppm
Identification (by IR) Passes Test
Phosphate (PO₄) ≤ 1 ppm
Residue after Evaporation ≤ 5 ppm
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤ 0.003 %
Water (H₂O) ≤ 0.02 %
Trace Impurities - Aluminum (Al) ≤ 0.100 ppm
Trace Impurities - Barium (Ba) ≤ 1.000 ppm
Trace Impurities - Boron (B) ≤ 0.100 ppm
Trace Impurities - Cadmium (Cd) ≤ 1.000 ppm
Trace Impurities - Calcium (Ca) ≤ 0.100 ppm
Trace Impurities - Chromium (Cr) ≤ 0.100 ppm
Trace Impurities - Cobalt (Co) ≤ 0.100 ppm
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
Trace Impurities - Gallium (Ga) ≤ 0.500 ppm
Trace Impurities - Germanium (Ge) ≤ 1.000 ppm
Trace Impurities - Gold (Au) ≤ 0.100 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Lead (Pb) ≤ 0.100 ppm
Trace Impurities - Lithium (Li) ≤ 1.000 ppm
Trace Impurities - Magnesium (Mg) ≤ 0.100 ppm
Trace Impurities - Manganese (Mn) ≤ 0.100 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Trace Impurities - Potassium (K) ≤ 0.100 ppm
Trace Impurities - Silicon (Si) ≤ 1.000 ppm
Trace Impurities - Silver (Ag) ≤ 0.500 ppm
Trace Impurities - Sodium (Na) ≤ 0.100 ppm
Trace Impurities - Strontium (Sr) ≤ 1.000 ppm
Trace Impurities - Tin (Sn) ≤ 0.100 ppm
Trace Impurities - Zinc (Zn) ≤ 0.100 ppm
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR