Zinc chloride ≥97%, granules, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
Management of Change (MOC) category = N
8780-04
8780-20
MK878006CS
3680.37
USD
MK878006
MK878004
MK878020
Zinc chloride ≥97%, granules, AR® ACS, Macron Fine Chemicals™
Zinc chloride
Granular.
Formula:
ZnCl₂ Boiling Pt: 732 °C (1013 hPa) Melting Pt: 283 °C Density: 2.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (ZnCl₂) | ≥97.0 % |
Ammonium (NH₄) | ≤0.005 % |
Calcium (Ca) | ≤0.06 % |
Insoluble Matter | ≤0.005 % |
Trace Impurities - Iron (Fe) | ≤0.001 % |
Lead (Pb) | ≤0.005 % |
Magnesium (Mg) | ≤0.01 % |
Nitrate (NO₃) | ≤0.003 % |
Oxychloride | Passes Test |
Potassium (K) | ≤0.02 % |
Sodium (Na) | ≤0.05 % |
Solubility | Passes Test |
Sulfate (SO₄) | ≤0.01 % |
Appearance (off-white to gray granules) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...