Zinc acetate dihydrate ≥98%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Acetic acid zinc salt dihydrate, Zinc (II) acetate dihydrate, Zinc diacetate dihydrate

Management of Change (MOC) category = NI

8740-04 8740-06
MK874004CS 1691.47 USD
MK874004 MK874006
Zinc acetate dihydrate ≥98%, granular, AR® ACS, Macron Fine Chemicals™
Zinc acetate dihydrate

Granular.

Formula: (H₃CCOO)₂Zn·2H₂O
Melting Pt: 237 °C
Density: 1.718 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((CH₃COO)₂Zn·2H₂O) 98.0 - 101.0 %
Solubility Passes Test
ACS - Chloride (Cl) ≤0.0005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iron (Fe) ≤0.0005 %
ACS - Lead (Pb) ≤0.002 %
ACS - Sulfate (SO₄) ≤0.002 %
ACS - pH of 5% Solution at 25°C 6.0 - 7.0
ACS - Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR