Xylene (mixture of isomers) ≥98.5% (by GC), AR® (ACS) Reagent Solvents ACS for histology, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Dimethylbenzene
Management of Change (MOC) category = N
8668-06
8668-22
8668-26
MK866806CS
658.26
USD
MK866806
MK866802
MK866822
MK866826
MK866816
Xylene (mixture of isomers) ≥98.5% (by GC), AR® (ACS) Reagent Solvents ACS for histology, Macron Fine Chemicals™
Xylene (mixture of isomers)
ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.
ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.
Formula:
C₆H₄(CH₃)₂ Boiling Pt: 140 °C (1013 hPa) Melting Pt: –34 °C Density: 0.86…0.88 g/cm³ (20 °C) Flash Pt: 29 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077264 CAS Number: 1330-20-7 EINECS: 215-535-7 UN: 1307 ADR: 3,III Merck Index: 13,10136 |
|
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (m,p,o-xylenes and ethylbenzene) (by GC) | ≥ 98.5 % |
ACS - Color (APHA) | ≤ 10 |
ACS - Ethylbenzene (C₆H₅C₂H₅) | ≤ 25 % |
Particulate Contamination | Passes Test |
ACS - Residue after Evaporation | ≤ 0.002 % |
ACS - Substances Darkened by H₂SO₄ | Passes Test |
Sulfur Compounds (as S) | ≤ 0.0005 % |
ACS - Water (H₂O) | ≤ 0.02 % |
Appearance (clear, colorless liquid) | |
Date Received: / / | |
Date Opened: / / |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...