D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERF MAT - MACRON LAB
Warning

Synonyms: alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar

Management of Change (MOC) category = N

8360-06 8360-24 8360-20
MK836006CS 1410.65 USD
MK836006 MK836024 MK836020
D-(+)-Sucrose, crystals, AR® ACS, Macron Fine Chemicals™
D(+)-Sucrose

Crystals.

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Solubility Passes Test
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
ACS - Titrable Acid (meq/g) ≤0.0008
ACS - Invert Sugar ≤0.05 %
ACS - Loss on Drying at 105°C ≤0.03 %
ACS - Residue after Ignition ≤0.01 %
ACS - Sulfate and Sulfite (as SO₄) ≤0.005 %
ACS - Specific Rotation [Å]²⁵^D (+) 66.3 - 66.8 Degree

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR