Sodium hydroxide ≥95%, pellets, GenAR® NF, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = R
7772-06
7772-25
7772-26
7772-88
7772-28
MACR7772-30
MK777206CS
915.63
USD
MK777206
MK777225
MK777226
MK777288
MK777228
JT7772-30
MK7772-30
Sodium hydroxide ≥95%, pellets, GenAR® NF, Macron Fine Chemicals™
Sodium hydroxide
Pellets. For biotechnology applications. GMP manufactured.
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 323 °C Density: 2.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Specification Test Results
Meets F.C.C. Requirements | |
GMP Manufactured Product | |
Meets E.P. Chemical Specifications | |
Meets N.F. Requirements | |
Food GMP Manufactured Product | |
Meets J.P. Chemical Specifications | |
Meets B.P. Chemical Specifications | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
NF - Total Alkali | 95.0 - 100.5 % |
NF - Content of Sodium | 54.0 - 59.8 % |
NF - Insoluble Substances and Organic Matter | Passes Test |
NF - Identification A (Sodium) | Passes Test |
NF - Identification B (pH) | ≥11.0 |
NF - Potassium (K) | ≤0.5 % |
NF/FCC - Carbonate (as Na₂CO₃) | ≤3.0 % |
FCC - Assay (Total alkali as NaOH) | 95.0 - 100.5 % |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Mercury (Hg)(mg/kg) | ≤0.1 |
FCC - Arsenic (As)(mg/kg) | ≤3 |
FCC - Identification | Passes Test |
FCC - Insoluble Substances and Organic Matter | Passes Test |
BP/EP - Assay (Total alkali as NaOH) | 97.0 - 100.5 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Iron (Fe) | ≤10 ppm |
EP/BP - Carbonate (as Na₂CO₃) | ≤2.0 % |
EP/BP - Chloride (Cl) | ≤200 ppm |
EP/BP - Sulfate (SO₄) | ≤200 ppm |
JP - Assay | 95.0 - 101.0 % |
JP - Identification A | Passes Test |
JP - Identification B | Passes Test |
JP - Clarity and Color of Solution | Passes Test |
JP - Chloride (Cl) | ≤0.050 % |
JP - Heavy Metals (as Pb) | ≤30 ppm |
JP - Potassium (K) | Passes Test |
JP - Carbonate (as Na₂CO₃) | ≤2.0 % |
JP - Mercury (Hg) | Passes Test |
Endotoxin Concentration (EU/g) | ≤5 |
Appearance (white hygroscopic pellets) | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...