Sodium hydroxide ≥95%, pellets, GenAR® NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Management of Change (MOC) category = R

7772-06 7772-25 7772-26 7772-88 7772-28 MACR7772-30
MK777206CS 915.63 USD
MK777206 MK777225 MK777226 MK777288 MK777228 JT7772-30 MK7772-30
Sodium hydroxide ≥95%, pellets, GenAR® NF, Macron Fine Chemicals™
Sodium hydroxide

Pellets. For biotechnology applications. GMP manufactured.

Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Meets F.C.C. Requirements
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Food GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
Bulk Food Chemical
NF - Total Alkali 95.0 - 100.5 %
NF - Content of Sodium 54.0 - 59.8 %
NF - Insoluble Substances and Organic Matter Passes Test
NF - Identification A (Sodium) Passes Test
NF - Identification B (pH) ≥11.0
NF - Potassium (K) ≤0.5 %
NF/FCC - Carbonate (as Na₂CO₃) ≤3.0 %
FCC - Assay (Total alkali as NaOH) 95.0 - 100.5 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Mercury (Hg)(mg/kg) ≤0.1
FCC - Arsenic (As)(mg/kg) ≤3
FCC - Identification Passes Test
FCC - Insoluble Substances and Organic Matter Passes Test
BP/EP - Assay (Total alkali as NaOH) 97.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Carbonate (as Na₂CO₃) ≤2.0 %
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤200 ppm
JP - Assay 95.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.050 %
JP - Heavy Metals (as Pb) ≤30 ppm
JP - Potassium (K) Passes Test
JP - Carbonate (as Na₂CO₃) ≤2.0 %
JP - Mercury (Hg) Passes Test
Endotoxin Concentration (EU/g) ≤5
Appearance (white hygroscopic pellets)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR