D-(+)-Sucrose, GenAR® NF for biotechnology, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = R

7723-88 7723-24 7723-04
MK772388EA 812.03 USD
MK772388 MK772324 MK772304
D-(+)-Sucrose, GenAR® NF for biotechnology, Macron Fine Chemicals™
D(+)-Sucrose

Crystals. For use in biotechnology. GMP manufactured.

Formula: C₁₂H₂₂O₁₁
Melting Pt: 169…170 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets N.F. Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
GMP Manufactured Product
Store in a tightly closed container.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Appearance of Solution Passes Test
NF - Color Value ≤45
NF - Conductivity ≤35 µS/cm
NF - Optical Rotation (+) 66.3 - 67.0 Degree
NF - Reducing Sugars Passes Test
NF - Sulfite (SO2) ≤10 ppm
NF - Loss on Drying at 105 ºC ≤0.1 %
EP/BP - Identification A Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Colour Value ≤45
EP/BP - Conductivity, µS cm⁻¹ ≤35
EP/BP - Optical Rotation (+) 66.3 - 67.0 Degree
EP/BP - Sulfite (SO₂) ≤10 ppm
EP/BP - Loss on Drying at 105°C ≤0.1 %
Endotoxin Concentration (EU/g) ≤250
EP/BP - Reducing Sugars Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR