Sodium hydroxide ≥95%, pellets NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERF MAT - MACRON LAB
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Management of Change (MOC) category = R
7680-06
7680-04
7680-20
7680-28
7680-88
MACR7680-25
MK768006CS
724.88
USD
MK768006
MK768004
MK768020
MK768028
MK768088
JT7680-QM
MK7680-26
MK7680-25
Sodium hydroxide ≥95%, pellets NF, FCC, Macron Fine Chemicals™
Sodium hydroxide
Pellets. GMP manufactured.
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 323 °C Density: 2.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Specification Test Results
GMP Manufactured Product | |
Food GMP Manufactured Product | |
Meets F.C.C. Requirements | |
Meets N.F. Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Bulk Food Chemical | |
NF - Total Alkali | 95.0 - 100.5 % |
NF - Content of Sodium | 54.0 - 59.8 % |
NF - Appearance (white hygroscopic pellets) | Passes Test |
NF - Insoluble Substances and Organic Matter | Passes Test |
NF - Identification A (Sodium) | Passes Test |
NF - Identification B (pH) | ≥11.0 |
NF - Potassium (K) | ≤0.5 % |
NF - Sodium Carbonate (Na₂CO₃) | ≤3.0 % |
FCC - Assay (Total alkali as NaOH) (by acidimetry) | 95.0 - 100.5 % |
FCC - Identification | Passes Test |
FCC - Insoluble Substances and Organic Matter | Passes Test |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Mercury (Hg) | ≤0.1 mg/kg |
FCC - Arsenic (As) | ≤3 mg/kg |
FCC - Sodium Carbonate (Na₂CO₃) | ≤3.0 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232) - Information on elemental impurities for | |
this product is available on the associated Product Regulatory Data | |
Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...