EDTA disodium salt dihydrate, GenAR® USP, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = R

MACR7565-21
JT7565-21CS 1369.74 USD
JT7565-21 MK7565-21
EDTA disodium salt dihydrate, GenAR® USP, Macron Fine Chemicals™
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (Dried Basis) 99.0 - 101.0 %
Appearance (white crystals or white powder) Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Identification C Passes Test
USP - pH (1 in 20) 4.0 - 6.0
USP - Loss on Drying 8.7 - 11.4 %
USP - Calcium (Ca) Passes Test
USP - Limit of Nitrilotriacetic Acid ≤0.1 %
EP - Assay 98.5 - 101.0 %
EP - Appearance of Solution Passes Test
EP - Identification A Passes Test
EP - Identification B Passes Test
EP - Identification D Passes Test
EP - pH.... 4.0 - 5.5
EP - Iron (Fe) ≤80 ppm
EP - Impurity A ≤0.1 %
JP - Assay 99.0 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Identification C Passes Test
JP - pH 4.3 - 4.7
JP - Clarity and Color of Solution Passes Test
JP - Cyanide (CN) Passes Test
JP - Heavy Metals ≤10 ppm
JP - Arsenic (As) ≤2 ppm
JP - Residue on Ignition 37.0 - 39.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR